Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAG5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521434
Description
Recommended positive controls: 293T, HeLa, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (92%), Rat (91%), Chicken (82%), Chimpanzee (99%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BAG5 Polyclonal antibody specifically detects BAG5 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
BAG5 | |
Polyclonal | |
Unconjugated | |
BAG5 | |
1600025G07Rik; 4930405J06Rik; BAG cochaperone 5; BAG family molecular chaperone regulator 5; Bag5; BAG-5; BAG-family molecular chaperone regulator-5; BAG-family molecular chaperone regulator-5 (BAG-5); BCL2 associated athanogene 5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5; KIAA0873; LOC522854 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
70369, 9529 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.68 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8CI32, Q9UL15 | |
BAG5 | |
Recombinant fragment corresponding to a region within amino acids 1 and 195 of BAG5 (Uniprot ID#Q9UL15) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction