Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAG5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA112709
Description
BAG5 Polyclonal antibody specifically detects BAG5 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
BAG5 | |
Polyclonal | |
PBS with 0.08% sodium azide | |
Q5QJC9, Q9UL15 | |
BAG5 | |
Synthetic peptide corresponding to human BAG-5. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
BAG5 | |
1600025G07Rik; 4930405J06Rik; BAG cochaperone 5; BAG family molecular chaperone regulator 5; Bag5; BAG-5; BAG-family molecular chaperone regulator-5; BAG-family molecular chaperone regulator-5 (BAG-5); BCL2 associated athanogene 5; Bcl-2-associated athanogene 5; BCL2-associated athanogene 5; KIAA0873; LOC522854 protein | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
366734, 9529 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction