Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI1 Isoform 3 Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131418

Catalog No. PA131418


Add to cart

Description

Description

PA1-31418 detects BAI1 protein 2 isoform 3 from human samples. PA1-31418 has been successfully used in Western blot applications, where a band of approximately 50 kDa is detected. Overnight incubation is recommended. Positive control: HeLa cell lysates. The PA1-31418 immunogen is a synthetic peptide containing residues SMSSADVEVARF. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

BAI 1 is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain.
Specifications

Specifications

BAI1 Isoform 3
Polyclonal
Unconjugated
ADGRB1
Liquid
ADGRB1
synthetic peptide SMSSADVEVARF.
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O14514
FLJ41988, GDAIF
Goat
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
575
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit