Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAIAP2L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522026
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (94%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BAIAP2L1 Polyclonal antibody specifically detects BAIAP2L1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
BAIAP2L1 | |
Polyclonal | |
Unconjugated | |
BAIAP2L1 | |
1300006M19Rik; AI585895; BAI1 associated protein 2 like 1; BAI1-associated protein 2-like 1; BAI1-associated protein 2-like protein 1; Baiap2l1; Brain-specific angiogenesis inhibitor 1-associated protein 2-like protein 1; insulin receptor tyrosine kinase substrate; IRTKS; RGD1308452 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.55 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9UHR4 | |
BAIAP2L1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 266 of BAIAP2L1 (Uniprot ID#Q9UHR4) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction