Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BAP1, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP126627

Catalog No. NBP126627

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BAP1
1.0 mg/mL
Immunoprecipitation 5-10 ug/mg lysate, Western Blot (Negative)
NP_004647.1
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Rabbit
IgG
100 ul
Apoptosis, Cancer, Cell Cycle and Replication, DNA Repair, Tumor Suppressors
Primary
8314
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
BAP1
The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human BRCA1-associated protein 1 using the numbering given in entry NP_004647.1 (GeneID 8314).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only