Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLVCR2 Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112457

Catalog No. 01-674-145


Add to cart

Description

Description

BAPTA ((1,2-bis(2-Aminophenoxy)ethane-N,N,N ft.,N ft.- tetraacetic acid) is a highly selective calcium chelator. BAPTA has spawned many of the most familiar fluorescent indicators for calcium and other ions. BAPTA and its analogs including BAPTA-AM have proven most critical in cell physiology research where they have been essential in defining the mechanisms underlying inter- or intracellular calcium homeostasis and calcium dependent exocytosis. BAPTA and its analogs may also find application in the study of diseases in which the regulation of excitatory neurotransmitter release and uptake has been disturbed.
Specifications

Specifications

FLVCR2
Polyclonal
PBS with 0.08% sodium azide
P60815
FLVCR2
Synthetic peptide derived from human BAPTA
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Rat
Western Blot
Unconjugated
Flvcr2
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
314323
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit