Learn More
BARHL1 Mouse anti-Human, Clone: 1B7, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522294
Catalog No. PIMA522294
Add to cart
Description
Description
Peptide Sequence: GRDCLETGTP RPGGASGPGL DSHLQPGQLS APAQSRTVTS SFLIRDILAD CKPLAACAPY SSSGQPAAPE PGGRLAAKAA EDFRDKLDKS GSNASSDSEY
BarH-like 1
Specifications
Specifications
BARHL1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9BZE3
PKCdeltaVIII
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56751
ELISA, Western Blot
1B7
Unconjugated
BARHL1
Liquid
BARHL1
BARHL1 (NP_064448, 55 a.a. ∽ 154 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human