Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BarH-like homeobox 1, Mouse, Clone: 2B11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BARHL1.
Supplier: Abnova Corporation H00056751M01
Description
Sequence: GRDCLETGTPRPGGASGPGLDSHLQPGQLSAPAQSRTVTSSFLIRDILADCKPLAACAPYSSSGQPAAPEPGGRLAAKAAEDFRDKLDKSGSNASSDSEYSpecifications
| BarH-like homeobox 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_020064 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 2B11 | |
| Mouse monoclonal antibody raised against a partial recombinant BARHL1. | |
| BARHL1 | |
| BARHL1 | |
| BARHL1 (NP_064448, 55 a.a. ∼ 154 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 56751 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction