Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

basic FGF Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610073

 View more versions of this product

Catalog No. BDB610073


Add to cart

Description

Description

Basic Fibroblast Growth Factor (bFGF) is a family member of the cell-differentiating and growth promoting factors. Basic FGF exists as multiple isoforms ranging from 18-24kDa that are derived from a single mRNA species under unique translational conditions. The 18kDa isoform is primarily cytosolic, but is also secreted and may form reservoirs of bFGF in the extracellular matrix. In contrast, the larger isoforms are predominantly nuclear. At the cellular level, bFGF is a potent mitogen and promotes cell survival by inhibiting apoptosis. At the tissue level, it is involved in wound repair and induces angiogenesis. In addition, bFGF is a significant target in cancer research because it is over-expressed in some cancers and may enhance a tumor's metastatic potential.

Host Species: Mouse
Clone: 6
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human bFGF aa. 1-155
Formula Weight [Chemical]: 18-24kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

basic FGF
6
Unconjugated
Affinity Purified
Human bFGF aa. 1-155
150μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.