Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG6, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127492

Catalog No. PIPA127492

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Unknown. A cluster of genes. BAT1-BAT5. has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is a nuclear prote

PA1-27492 detects BAT3 from human and rat samples.PA1-27492 may be successfully used in Western blot applications.The PA1-27962 immunogen is a synthetic peptide corresponding to human Large proline-rich protein BAT3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BAT3
Polyclonal
Unconjugated
P46379, Q6MG49
BAG6
Synthetic peptide corresponding to human Large proline-rich protein BAT3.
50 μg
Primary
Human, Rat
Antibody
IgY
Western Blot
1mg/mL
BAG6
DAQB-195H10.3, BAG-6, BAG6, D6S52E, G3
Chicken
Antigen affinity chromatography
RUO
7917
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit