Learn More
Bax Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB2915
Description
This Anti-Bax Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Bax.
Specifications
Bax | |
Polyclonal | |
Liquid antiserum, no preservatives. | |
Rabbit | |
100 μL | |
NM_004324.3; NM_138761.2; NM_138763.2; NM_138764.2; NM_138765.2 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
BAX | |
A synthetic peptide corresponding to amino acids 43-61 of the mouse Bax-alpha protein. | |
Primary | |
Mouse | |
Serum |