Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCL13 Mouse anti-Human, APC, Clone: 53610, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523629

Catalog No. PIMA523629


Add to cart

Description

Description

In ELISAs, this antibody does not cross-react with recombit human (rh) CXCL1, rhCXCL2, rhCXCL3, rhCXCL8/IL-8, rhCXCL10, rhCXCL9, rhCXCL12/SDF-1 alpha, or rhCXCL12/SDF-1 beta.

B lymphocyte chemoattractant, independently cloned and named Angie, is a CXC chemokine strongly expressed in the follicles of the spleen, lymph nodes, and Peyer ft.s patches. It preferentially promotes the migration of B lymphocytes (compared to T cells and macrophages), apparently by stimulating calcium influx into, and chemotaxis of, cells expressing Burkitt ft.s lymphoma receptor 1 (BLR-1). It may therefore function in the homing of B lymphocytes to follicles.
Specifications

Specifications

CXCL13
Monoclonal
APC
CXCL13
Liquid
CXCL13
E. coli-derived recombit human CXCL13/BLC/BCA-1 Val23-Arg94
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
53610
Buffered saline solution with BSA and 0.09% sodium azide
O43927
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13, B-cell chemoattractant, B-cell-attracting chemokine 1, B-cell-homing chemokine (ligand for Burkitt′s lymphoma receptor-1), B-lymphocyte chemoattractant, C-X-C motif chemokine 13, CXC chemokine BLC, b cell-attracting chemokine 1, b lymphocyte chemoattractant, chem
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
10563
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit