Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CXCL13 Monoclonal Antibody (53610), PE, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523666
Description
In ELISAs, this antibody does not cross-react with recombit human (rh) CXCL1, rhCXCL2, rhCXCL3, rhCXCL8/IL-8, rhCXCL10, rhCXCL9, rhCXCL12/SDF-1 alpha, or rhCXCL12/SDF-1 beta.
B lymphocyte chemoattractant, independently cloned and named Angie, is a CXC chemokine strongly expressed in the follicles of the spleen, lymph nodes, and Peyer′s patches. It preferentially promotes the migration of B lymphocytes (compared to T cells and macrophages), apparently by stimulating calcium influx into, and chemotaxis of, cells expressing Burkitt′s lymphoma receptor 1 (BLR-1). It may therefore function in the homing of B lymphocytes to follicles.Specifications
CXCL13 | |
Monoclonal | |
PE | |
CXCL13 | |
ANGIE, ANGIE2, BCA-1, BCA1, BLC, BLR1L, SCYB13, B-cell chemoattractant, B-cell-attracting chemokine 1, B-cell-homing chemokine (ligand for Burkitt′s lymphoma receptor-1), B-lymphocyte chemoattractant, C-X-C motif chemokine 13, CXC chemokine BLC, b cell-attracting chemokine 1, b lymphocyte chemoattractant, chem | |
Mouse | |
Protein A/G | |
RUO | |
10563 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
53610 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
O43927 | |
CXCL13 | |
E. coli-derived recombit human CXCL13/BLC/BCA-1 Val23-Arg94 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction