Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B-cell receptor-associated protein 29, Mouse, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human BCAP29 protein.
Manufacturer: Abnova Corporation H00055973B01
Description
Sequence: MTLQWAAVATFLYAEIGLILIFCLPFIPPQRWQKIFSFNVWGKIATFWNKAFLTIIILLIVLFLDAVREVRKYSSVHTIEKSSTSRPDAYEHTQMKLFRSQRNLYISGFSLFFWLVLRRLVTLITQLAKELSNKGVLKTQAENTNKAAKKFMEENEKLKRILKSHGKDEECVLEAENKKLVEDQEKLKTELRKTSDALSKAQNDVMEMKMQSERLSKEYDQLLKEHSELQDRLERGNKKRLSpecifications
B-cell receptor-associated protein 29 | |
Unconjugated | |
No additive | |
NM_018844 | |
BAP29|DKFZp686M2086 | |
Mouse | |
50 uL | |
Apoptosis | |
Y | |
Polyclonal | |
Human |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human BCAP29 protein. | |
BCAP29 | |
Serum | |
BCAP29 | |
BCAP29 (NP_061332, 1 a.a. ~ 241 a.a) full-length human protein. | |
RUO | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
55973 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title