Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAS2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA120841

Catalog No. PIPA120841

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1.5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.

PA1-20841 detects BCAS2 in human, mouse, rat and Xenopus laevis samples.PA1-20841 has been successfully used in Western blot procedures.The PA1-20841 immunogen is full length human BCAS2.
Specifications

Specifications

BCAS2
Polyclonal
Unconjugated
BCAS2
Purified
BCAS2, bcas2.L
Full length human BCAS2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat, Xenopus laevis
Immunohistochemistry, Western Blot, Western Blotting
1mg/mL
PBS with no preservative
O75934, Q9D287
DAM1, SPF27, Snt309
Chicken
IgY
50μg
Store at -20°C
Primary
10286
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit