Learn More
BRCA2 and CDKN1A interacting protein, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human BCCIP protein.
Manufacturer: Abnova Corporation H00056647B01
Description
This gene product was isolated on the basis of its interaction with BRCA2 and p21 proteins. It is an evolutionarily conserved nuclear protein with multiple interacting domains. The N-terminal half shares moderate homology with regions of calmodulin and M-calpain, suggesting that it may also bind calcium. Functional studies indicate that this protein may be an important cofactor for BRCA2 in tumor suppression, and a modulator of CDK2 kinase activity via p21. This protein has also been implicated in the regulation of BRCA2 and RAD51 nuclear focus formation, double-strand break-induced homologous recombination, and cell cycle progression. Multiple transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Sequence: MASRSKRRAVESGVPQPPDPPVQRDEEEEKEVENEDEDDDDSDKEKDEEDEVIDEEVNIEFEAYSLSDNDYDGIKKLLQQLFLKAPVNTAELTDLLIQQNHIGSVIKQTDVSEDSNDDMDEDEVFGFISLLNLTERKGTQCVEQIQELVLRFCEKNCEKSMVEQLDKFLNDTTKPVGLLLSERFINVPPQIALPMYQQLQKELAGAHRTNKPCGKCYFYLLISKTFVEAEKNNSKKKPSNKKKAALMFANAEEEFFYEKAILKFNYSVQEESDTCLGGKWSFDDVPMTPLRTVMLIPGDKMNEIMDKLKEYLSVSpecifications
BRCA2 and CDKN1A interacting protein | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human BCCIP protein. | |
BCCIP | |
TOK-1/TOK1 | |
Mouse | |
50 μL | |
Cell Cycle | |
Primary | |
Human |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
BC009771 | |
BCCIP | |
BCCIP (AAH09771, 1 a.a. ∼ 314 a.a) full-length human protein. | |
RUO | |
Yes | |
56647 | |
Serum |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.