Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112513

Catalog No. 01-674-173


Add to cart

Description

Description

A suggested positive control for this product is Raji whole cell lysate.

BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
Specifications

Specifications

BCL-10
Polyclonal
PBS with 0.02% sodium azide
O95999
B-cell CLL/lymphoma 10, Bcl-10 CED-3/ICH-1 prodomain homologous E10-like regulator, CIPER
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8915
Western Blot
Unconjugated
BCL10
Liquid
BCL10
Synthetic peptide corresponding to residues 5-19 of human BCL-10.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit