Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BCL-2α Ab-4, Clone: 10C4, Thermo Scientific Lab Vision

Mouse Monoclonal Antibody

Specifications

Antigen BCL-2α Ab-4
Host Species Murine
Immunogen A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein.
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS598B0
View Documents
Epredia
MS598B0
100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS598B1
View Documents
Epredia
MS598B1
500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS598B
View Documents
Epredia
MS598B
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse
Clone: 10C4
Isotype: IgG1
Cross Reactivity: Mouse and Rat. Does not react with human. Others-not tested.
Epitope: aa 61-76
Immunogen: A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein.
Molecular Weight: 25-26kDa
Positive Control: MAD109 cells.
Cellular Localization: Cytoplasm

Ab-4 is particularly designed for research on mouse and rat bcl-2alpha protein. It does not react with human bcl-2alpha protein.

Immunofluorescence, Immunoprecipitation, Western Blotting

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

BCL-2α Ab-4
A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein.
Monoclonal
Murine
IgG1
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit