Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141087

Catalog No. 01-676-678


Add to cart

Description

Description

Suggested positive control: Jurkat whole cell lysate.

NF-kB is silenced in the cytoplasm by an inhibitory protein, IkB (1). The proto-oncoprotein Bcl-3 is a member of the IkB family and is present predomitly in the nucleus. IkBa and Bcl-3 are similar in the central ankyrin repeat domains, where as they differ at the N- and –C terminus. These differences may explain the differential binding of IkBa and Bcl-3. Two molecules of Bcl-3 can associate with NF-kB, where as only one molecule of IkBa bind to NF-kB. BCL-3 mRNA and protein expression are induced by NF-kB-activating agents. Down-regulation of BCL-3 induction results in prolonged, enhanced NF-kB binding and increased NF-kB-dependent transcription. Several cytokines, such as, TNFa or IL-1b, and mitogens as phorbolester (PMA) trigger rapid formation of Bcl-3-p50 complexes with the same kinetics as activation of p50-p65 complexes.
Specifications

Specifications

BCL-3
Polyclonal
Unconjugated
BCL3
Liquid
BCL3
Synthetic peptides corresponding to residues 2-13 and 107-120 of human Bcl-3.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P20749
BCL4, D19S37
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
602
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit