Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520234

Catalog No. PIPA520234


Add to cart

Description

Description

Despite its predicted molecular weight, Bcl-B often migrates at higher molecular weights, presumably due to post-translational modifications. A suggested positive control is Jurkat cell lysate. PA5-20234 can be used with blocking peptide PEP-0353.

Members in the Bcl-2 family are critical regulators of apoptosis by either inhibiting or promoting cell death. Bcl-B is a recently discovered anti-apoptotic member of the Bcl-2. Unlike the mouse homolog (also known as Diva/Boo) which is predomitly expressed in ovary and testis, the human Bcl-B protein is widely expressed. Also, the human Bcl-B functions by binding to and suppressing the apoptotic activity of Bax, whereas the mouse homolog binds Bak and also interacts with the apoptosis protein Apaf-1.
Specifications

Specifications

Bcl-B
Polyclonal
Unconjugated
BCL2L10
Liquid
BCL2L10
A 16 amino acid peptide from near the amino terminus of human Bcl-B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9HD36
BCL-B, Boo, Diva; anti-apoptotic protein NrH; apoptosis regulator Bcl-B; bcl-2-like protein 10; bcl2-L-10
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
10017
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit