Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-G Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA146277

Catalog No. PIPA146277

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Suggested positive control: antigen standard for BCL2L14 (transient overexpression lysate).

Bcl-G is a novel pro-apoptotic member of the Bcl-2 family. The human Bcl-G gene encodes two proteins through alternative mRNA splicing, Bcl-G(L) (long) and Bcl-G(S) (short) consisting of 327 and 252 amino acids in length, respectively. Bcl-G(L) mRNA is expressed widely in adult human tissues, whereas Bcl-G(S) mRNA was found only in testis. Overexpression of Bcl-G(L) or Bcl-G(S) in cells induced apoptosis although Bcl-G(S) was far more potent than Bcl-G(L).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BCL-G
Polyclonal
Unconjugated
Q9BZR8
BCL2L14
Residues 298-312.
100 μg
Primary
Human
Antibody
IgG
Western Blot
1 mg/mL
BCL2L14
BCLG
Rabbit
Antigen affinity chromatography
RUO
79370
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit