Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-X Mouse, Unlabeled, Clone: 4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610209

 View more versions of this product

Catalog No. BDB610209


Add to cart

Description

Description

Bcl-x is related to the Bcl-2 protein and can function independently of Bcl-2 in regulating apoptosis (programmed cell death). The bcl-x transcript is found in a number of tissues, with the highest levels in the lymphoid and central nervous systems. Two distinct cDNA species, bcl-xL and bcl-xS, have been observed and appear to arise from alternate 5 ft. splice sites located within the first coding exon of the bcl-x gene. Bcl-xL is composed of 233 amino acids and is similar in size and structure to Bcl-2. Bcl-xS lacks 63 amino acids corresponding to the region of Bcl-xL with the highest degree of homology to Bcl-2. Like Bcl-2, Bcl-xL has been reported to inhibit cell death upon growth factor withdrawal in an IL-3-dependent cell line. However, Bcl-xS inhibits the ability of Bcl-2 to enhance cell survival in the absence of growth factors.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Bcl-X
4
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
50μg
Cell Biology
Primary
Chicken, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.