Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-xL Mouse anti-Human, Mouse, Clone: 7D9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA137166

Catalog No. 01-671-678


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is Hodgkin ft.s lymphoma.

BCL-XL is a part of the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BCL-XL is located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential and controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator.
Specifications

Specifications

BCL-XL
Monoclonal
Unconjugated
BCL2L1
Liquid
BCL2L1
Synthetic peptide corresponding to N-terminus of mouse BCL-XL protein
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse
Immunohistochemistry (Paraffin)
7D9
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Q07817, Q64373
BCL-XL/S, BCL2L, BCLX, Bcl-X, DKFZp781P2092, bcl-xL, bcl-xS
Mouse
IgG2a
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12048, 598
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit