Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-xL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141101

Catalog No. 01-675-690


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology. Suggested positive control: antigen standard for BCL2L1 (transient overexpression lysate), Jurkat, MOLT-4, or Rh-30.

BCL-XL is a part of the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BCL-XL is located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential and controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported. The longer isoform acts as an apoptotic inhibitor and the shorter form acts as an apoptotic activator.
Specifications

Specifications

BCL-XL
Polyclonal
Unconjugated
BCL2L1
Liquid
BCL2L1
Synthetic peptide corresponding to residues 3-14 of human and mouse Bcl-XL and Bcl-XS.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
1.0 mg/mL
PBS with 0.02% sodium azide
Q07817
BCL-XL/S, BCL2L, BCLX, Bcl-X, DKFZp781P2092, bcl-xL, bcl-xS
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
598
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit