Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-rambo Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515042

Catalog No. PIPA515042

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

BCL2L10 belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein may promote the activation of caspase-3 and apoptosis.
Specifications

Specifications

Bcl-rambo
Polyclonal
PBS with 0.09% sodium azide
Q9BXK5
Bcl-rambo; Protein Mil1; Bcl-2-like 13 protein; BCL2L13; MIL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23786
Western Blot
Unconjugated
BCL2L13
Liquid
BCL2L13
KLH conjugated synthetic peptide between 342-369 amino acids from the C-terminal region of human BCL2L13
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit