Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDH Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525466
Description
This antibody is predicted to react with bovine, mouse, porcine, non-human primate and rat based on sequence homology.
The pyruvate dehydrogenase (PDH) complex is a nuclear-encoded mitochondrial multienzyme complex that catalyzes the overall conversion of pyruvate to acetyl-CoA and CO(2), and provides the primary link between glycolysis and the tricarboxylic acid (TCA) cycle. The PDH complex is composed of multiple copies of three enzymatic components: pyruvate dehydrogenase (E1), dihydrolipoamide acetyltransferase (E2) and lipoamide dehydrogenase (E3). The E1 enzyme is a heterotetramer of two alpha and two beta subunits. This gene encodes the E1 alpha 1 subunit containing the E1 active site, and plays a key role in the function of the PDH complex.Specifications
PDH | |
Polyclonal | |
PDHA1 | |
PDHA, PDHCE1A, PHE1A, PDHE1-A type I, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial, pyruvate dehydrogenase complex, E1-alpha polypeptide 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5160 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
P08559 | |
PDHA1 | |
KLH conjugated synthetic peptide between 228-255 amino acids from the central region of human PDHA1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction