Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BDNF Monoclonal Antibody (35909), APC, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523665
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) NT-3, rhNT-4, or rh beta -NGF is observed.
BDNF Monoclonal antibody specifically detects BDNF in Human samples. It is validated for Flow Cytometry.Specifications
BDNF | |
Monoclonal | |
APC | |
BDNF | |
abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
Mouse | |
Protein A/G | |
RUO | |
627 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
35909 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P23560 | |
BDNF | |
Sf 21-derived recombinant human BDNF His129-Arg247 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction