Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BDNF, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP141436

Catalog No. NBP141436

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BDNF
LYOPH
Western Blot 1-2 ug/ml
Affinity Purified
BDNF
A peptide mapping at the middle region of human BDNF, identical to the related rat and mouse sequence.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Reconstitute with 0.1 ml deionized water.
Western Blot
Unconjugated
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3. with 0.05mg Thimerosal
abrineurin, brain-derived neurotrophic factor, MGC34632, neurotrophin
Rabbit
IgG
0.1 mg
Cytokine Research, Immunology, Neuroscience
Primary
627
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only