Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Bestrophin, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB5466/DEL

Description
Immunohistochemistry, Western Blotting

Specifications
Bestrophin | |
Unconjugated | |
Recombinant human bestrophin protein. | |
RUO | |
Primary | |
Semi-purified |
Monoclonal | |
Mouse | |
100 μg | |
Neuroscience | |
Human, Monkey, Pig | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction