Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-2 Adrenergic Receptor, Polyclonal, Invitrogen™

Catalog No. PIPA125659
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA125659 50 μg
1 options

Catalog No. PIPA125659

Supplier: Thermo Scientific PA125659

Chicken Polyclonal Antibody

This gene encodes beta-2-adrenergic receptor which is a member of the G protein-coupled receptor superfamily. This receptor is directly associated with one of its ultimate effectors, the class C L-type calcium channel Ca(V)1.2. This receptor-channel complex also contains a G protein, an adenylyl cyclase, cAMP-dependent kinase, and the counterbalancing phosphatase, PP2A. The assembly of the signaling complex provides a mechanism that ensures specific and rapid signaling by this G protein-coupled receptor. This gene is intronless. Different polymorphic forms, point mutations, and/or downregulation of this gene are associated with nocturnal asthma, obesity and type 2 diabetes.

PA1-25659 detects beta 2 Adrenergic Receptor from human samples.PA1-25659 has been successfully used in Western blot procedures.The PA1-25659 immunogen is a synthetic peptide corresponding to residues 335-408 of human beta 2 Adrenergic Receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-2 Adrenergic Receptor
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene ADRB2
Gene Accession No. P07550
Gene Alias ADRB2R, ADRBR, B2AR, BAR, BETA2AR
Gene Symbols ADRB2
Host Species Chicken
Immunogen Synthetic peptide corresponding to residues 335-408 of human beta 2 Adrenergic Receptor.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.