Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-3 Defensin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA14216
Description
PA1-4216 detects the beta-Defensin-3 protein from human samples. PA1-4216 has been successfully used in Western blot and immunohistochemistry procedures. The PA1-4216 immunogen is a synthetic peptide corresponding to residues G(23) I I N T L Q K Y Y C(33) of the 5 kDa human beta Defensin-3 protein.
beta-3 Defensin Polyclonal antibody specifically detects beta-3 Defensin in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
beta-3 Defensin | |
Polyclonal | |
Unconjugated | |
DEFB103A | |
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
414325 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P81534 | |
DEFB103A | |
Synthetic peptide corresponding to residues G(23) I I N T L Q K Y Y C(33) of human beta Defensin-3. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction