Learn More
UbpbioSupplier Diversity Partner ANTI-BETA-ACTIN, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1051
Antibodies – Anti-beta-Actin (UBPBio catalog# Y1051) – Mouse monoclonal antibody (clone BA3R, IgG2b). Used for immunoblotting at 1:1,000-5,000 dilution for detection of beta-actin in 20 ug whole cell lysates. Recognizing beta-actin in human, mouse, rat, chicken and rabbit.
Catalog No. 50-112-6920