Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Actin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121167

Catalog No. PA121167

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Beta actin is one of six different actin isoforms which have been identified. Actins are highly conserved proteins that are involved in cell motility, structure and integrity.

PA1-21167 detects beta-actin in human, mouse, rat, bovine, sheep, rabbit, feline, chicken, canine, porcine, Hirudo medicinalis (leech), Drosophila melanogaster, and guinea pig samples.PA1-21167 has been successfully used in immunohistochemistry (FFPE) and Western blot procedures. Formalin-fixed paraffin-embedded tissue sections require boiling in 10 mM citrate buffer (pH 6.0) prior to immunostaining. The PA1-21167 immunogen is a synthetic peptide corresponding to residues A(26) P R A V F P S(33) of beta actin.
Specifications

Specifications

beta Actin
Polyclonal
Unconjugated
ACTB
Purified
ACTB
Synthetic peptide corresponding to residues A(26) P R A V F P S(33) of beta actin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Many
Immunohistochemistry (Paraffin), Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P60712, O18840, P60706, P60708, Q71FK5, P60709, P60710, P60713, Q6QAQ1, P29751, P60711
PS1TP5BP1, Atcb
Rabbit
IgG
500μL
Store at 4°C, do not freeze
Primary
60
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit