Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-beta Amyloid, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP135302

Catalog No. NBP135302


Add to cart

Description

Description

beta Amyloid Polyclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

beta Amyloid
Unconjugated
50mM Tris (pH 7.6) and 1.0% BSA with 0.1% Sodium Azide
Abeta, Amyloid beta, beta-amyloid peptide
Rabbit
IgG
7 ml
Store at 4C. Do not freeze.
Polyclonal
This product is specific for Amyloid b/Amyloid A4
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin Neat
Affinity Purified
APP
A synthetic peptide from N-terminus of human amyloid beta.
Immunogen affinity purified
RUO
Primary
351
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only