Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta Catenin Mouse anti-Human, Mouse, Rat, Clone: 7F7.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04958
Description
This Anti-Beta Catenin Antibody, clone 7F7.2 is validated for use in western blot, IC for the detection of Beta Catenin.Specifications
Beta Catenin | |
Monoclonal | |
Unconjugated | |
NP_001895 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Western Blot | |
7F7.2 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine, pH 7.4, 150mM NaCl with 0.05% sodium azide. | |
CTNNB, CTNNB1, FLJ25606, FLJ37923, DKFZp686D02253 | |
Full length, GST-tagged recombinant beta catenin. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only