Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCNN1B Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA1921

Catalog No. PA1921


Add to cart

Description

Description

PA1-921 detects recombit rat beta-epithelial sodium channel (beta-ENaC). PA1-921 has been successfully used in immunoprecipitation procedures. By immunoprecipitation this antibody specifically binds to the unglycosylated, ~75 kDa recombit rat beta-ENaC protein. This product fails to detect beta-ENaC protein in Western blot procedures. PA1-921 immunizing peptide corresponds to amino acid residues 619-640 from human beta-ENaC. This sequence is 90% and 81% conserved in the rabbit, rat and mouse beta-ENaC, respectively. PA1-921 immunizing peptide (Cat. No. PEP-089) is available for use in neutralization and control experiments.

Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel’s syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
Specifications

Specifications

SCNN1B
Polyclonal
Unconjugated
Scnn1b
Liquid
SCNN1B
Synthetic peptide corresponding to residues N(619) Y D S L R L Q P L D V I E S D S E G D A I(640) of rat beta-ENaC
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Rat
Immunoprecipitation
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
P37090
Epithelial Sodium Channel Beta
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
24767
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit