Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Polyclonal Antibody, Invitrogen™

beta Galactosidase Antibody, PA1-21477, from Invitrogen™. Species Reactivity: Bacteria; Applications: Dot blot, ELISA, ICC, IF, IHC (Paraffin, Frozen), IP, WB.

Manufacturer:  Invitrogen PA121477

Catalog No. PA121477

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

beta Galactosidase Antibody, PA1-21477, from Invitrogen. Species Reactivity: Bacteria; Applications: Dot blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin, Frozen), Immunoprecipitation, Western Blot.

Beta galactosidase is coded by a gene (lac z) in the lac operon of Escherichia coli. It is a metalloenzyme that splits lactose into glucose and galactose. It hydrolyzes terminal, non-reducing beta-D-galactose residues in beta-D-galactosides. Activation by cations seems to be substrate dependent. K+, Na+, NH4+, Rb+, Cs+ and Mn++ all activate enzyme activity based upon the substrate used.
Host Species: Rabbit
Cross Reactivity: Bacteria
Isotype: IgG
Immunogen: Recombinant full length beta Galactosidase protein expressed in E. coli.
Formulation: 1mg purified antibody (10mg/mL) in 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Synonyms: Beta-Gal, RP3-337O18.1, GLB2, GSL, NGBE, PPCA, PPGB
PA1-21477 detects beta Galactosidase in E. coli samples. PA1-21477 has been successfully used in ELISA, immunocytochemistry, immunofluorescence, immunohistochemistry (frozen and FFPE), immunoprecipitation, dot blot and Western blot procedures. The PA1-21477 immunogen corresponds to recombinant full length beta Galactosidase protein expressed in E. coli. Store at 4ºC short term. For extended storage, store at -20ºC, avoiding freeze/thaw cycles.
Specifications

Specifications

beta Galactosidase
Polyclonal
1mg purified antibody (10mg/mL) in 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Beta-Gal, RP3-337O18.1, GLB2, GSL, NGBE, PPCA, PPGB
Recombinant full length beta Galactosidase protein expressed in E. coli.
1mg
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
Bacteria
Dot Blot, ELISA, Frozen), Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin/Frozen), Immunoprecipitation, Western Blot
Unconjugated
Purified
Rabbit
IgG
RUO
Primary
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit