Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AP3B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA11067

Catalog No. 01-670-135


Add to cart

Description

Description

PA1-1067 detects beta NAP from human, rat, and mouse samples. PA1-1067 has been successfully used in Western blot applications. By Western blot this antibody detects a ~119-130 kDa protein representing beta NAP from brain samples. The PA1-1067 immunogens is a mixture of syntehtic peptides corresponding to residues L(260) L E E N (A/P) E K A F Y G S E E(274) of human and mouse beta NAP. The immunizing peptides (Cat. No. PEP-294) are available for purchase for use in neutralization controls.

Beta NAP is closely related to the beta-adaptin and beta-COP coat proteins. Beta NAP is a nonclathrin-associated phosphoprotein expressed exclusively in neurons, from E12 through adulthood. Beta NAP is present in the neurnal soma and nerve terminal as soluble and membrane-bound pools and is associated with a discrete set of nerve-terminal vesicles.
Specifications

Specifications

AP3B2
Polyclonal
Unconjugated
AP3B2
Liquid
AP3B2
Syntehtic peptides corresponding to residues L(260) L E E N (A/P) E K A F Y G S E E(274) of human and mouse beta NAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
antibody with 1mg/mL BSA and 0.05% sodium azide
Q13367, Q9JME5
Adapter-related protein complex 3 beta-2 subunit; Beta3B-adaptin; Adaptor protein complex AP-3 beta-2 subunit; AP-3 complex beta-2 subunit; Clathrin a
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11775, 308777, 8120
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit