Learn More
NGF, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA118378
Catalog No. 01-670-630
Description
Description
Beta NGF is a nerve growth factor that is important for the development and maintenance of the sympathetic and sensory nervous systems. It stimulates division and differentiation of sympathetic and embryonic sensory neurons.
By ELISA, this antibody exhibits a cross-reactivity of less than 1% to recombinant human BDNF, NT3 and NT4/5. It will not react with bovine NGF.Use at 1-3μg/ml for inhibition of biological activity in vitro. Use neat for in vivo studies at 2-10μg/ml (ED50). This antiserum completely inhibits neuronal survival and the outgrowth actions of murine NGF in chicken DRG in vitro.Reconstitute with 20 ul of distilled water. After reconstitution keep aliquots at -20ºC for a higher stability, and at 4ºC with an appropriate antibacterial agent.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NGF
Polyclonal
Unconjugated
NGF
beta Nerve Growth Factor
Rabbit
Affinity chromatography
RUO
18049, 310738, 4803
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Lyophilized
ELISA, Immunohistochemistry, Inhibition Assays, Western Blot
1mg/mL
Antibody with no preservative
P01138, P01139, P25427
NGF
Native mouse beta NGF purified from submaxillary salivary gland.
20 μg
Primary
Avian, Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.