Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Synuclein Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138703

Catalog No. 01-674-692


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, canine, mouse, porcine, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain tissue.

This antibody is specific for beta-synuclein and does not cross react with alpha-and gamma-synuclein Synucleins are small, soluble proteins expressed primarily in neural tissue and in certain tumors. The family includes three known proteins: alpha-synuclein, beta-synuclein, and gamma-synuclein. The alpha- and beta-synuclein proteins are found primarily in brain tissue, where they are seen mainly in presynaptic terminals. The gamma-synuclein protein is found primarily in the peripheral nervous system and retina, but its expression in breast tumors makes it a candidate as a marker for tumor progression.
Specifications

Specifications

beta Synuclein
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
Q16143
beta-synuclein
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6620
Immunohistochemistry (Paraffin)
Unconjugated
SNCB
Liquid
SNCB
Synthetic peptide derived from near C-terminus of human synuclein-beta.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit