Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Tubulin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA121153

Catalog No. PA121153

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and in the cytoskeleton.

PA1-21153 detects beta-Tubulin in guinea pig, human, mouse, rat, fungus, S. cerevisiae, bovine, porcine, sea urchin, gerbil, and chicken samples.PA1-21153 has been successfully used in immunohistochemistry (paraffin), immunoprecipitation and Western blot procedures. Suggested positive controls for this product are MAD109 cells, skin and lung.The PA1-21153 immunogen is a synthetic peptide corresponding to residues N(416) D L V S E Y Q Q Y Q D A T A(430) of human beta Tubulin.
Specifications

Specifications

beta Tubulin
Polyclonal
Unconjugated
TUBB4A
Purified
TUBB, TUBB4A
Synthetic peptide corresponding to residues N(416) D L V S E Y Q Q Y Q D A T A(430) of human beta Tubulin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Many
Functional Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
Q3ZBU7, P04350, P07437, Q9D6F9, Q767L7
M40 antibody, Tubulin beta polypeptide antibody.
Rabbit
IgG
500μL
Store at 4°C, do not freeze
Primary
10382, 203068
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit