Learn More
NADE Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSB00009W
Catalog No. PIOSB00009W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
NADE
Polyclonal
whole serum with no preservative
Q9WTZ9, Q6PDU5
Protein BEX3, NGFRAP1, p75NTR-associated cell death executor, Nerve growth factor receptor-associated protein 1 (TNFRSF16) associated protein 1, Ovari
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Bex3
Lyophilized
Bex3
A synthetic peptide from n-terminal region of mouse Brain-expressed X-linked protein 3
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
117089, 12070