Learn More
TGR5 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523183
Catalog No. PIPA523183
Add to cart
Description
Description
BG37 is a 330 amino acid novel enzyme with seven transmembrane domains. It belongs to the G protein-coupled receptor (GPCR) superfamily and functions as a cell surface receptor for bile acids. Cells expressing BG37 when treated with bile acids induce the production of intracellular cAMP, activate the MAP kinase signaling pathway, leads to the internalization of the receptor and regulates proglucagon gene expression in enteroendocrine cells. The receptor is implicated in the suppression of macrophage functions and regulation of energy homeostasis by bile acids that can be targeted to improve metabolic control. It is known to be ubiquitously expressed in most of the tissues.
Specifications
Specifications
TGR5
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q8TDU6, Q80SS6, Q80T02
BG37, GPCR19, GPR131, M-BAR, TGR5, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, hBG37, hGPCR19, membrane bile acid receptor, membrane-type receptor for bile acids
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate, Rat
Western Blot
Unconjugated
GPBAR1
Liquid
GPBAR1
Synthetic peptide (41 WDRRLRSPPA 50) of BG37/TGR5 protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
151306, 227289, 338443