Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLH3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525651

Catalog No. PIPA525651


Add to cart

Description

Description

BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.
Specifications

Specifications

BHLH3
Polyclonal
PBS with 0.09% sodium azide
Q9C0J9
BHLHB3, DEC2, SHARP1, hDEC2, basic helix-loop-helix domain containing, class B, 3, class E basic helix-loop-helix protein 41, differentially expressed in chondrocytes 2, differentially expressed in chondrocytes protein 2, enhancer-of-split and hairy-related protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79365
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
BHLHE41
Liquid
BHLHE41
KLH conjugated synthetic peptide between 17-46 amino acids from the N-terminal region of human BHLH3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit