Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bid Mouse anti-Mouse, Unlabeled, Clone: 40, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611866

Catalog No. BDB611866


Add to cart

Description

Description

Members of the Bcl-2 protein family function to inhibit (Bcl-2, Bcl-XL, Mcl-1, A1) or promote (Bax, Bak, Bcl-Ls, Bad) apoptosis. Bid is a Bcl-2 family death agonist that heterodimerizes with either agonists (Bax) or antagonists (Bcl-2). Bid contains a centrally located BH3 domain that allows interaction with the BH1 domain of Bax and Bcl-2, as well as two caspase cleavage sites (CCS). However, Bid does not contain other domains commonly found in Bcl-2 family members, such as BH1, BH2, or BH4. In addition, Bid does not contain a C-terminal hydrophobic region that is characteristic of membrane-bound Bcl-2 family members. Singlet oxygen-induced apoptosis in human leukemia cells and Fas-induced apoptosis involve caspase-8 cleavage of Bid, which produces a 15 kDa C-terminal fragment and a 6.5 kDa N-terminal fragment. The C-terminal fragment translocates to the mitochondria and promotes the release of cytochrome C through a mechanism that might involve destabilization of the mitochondrial membrane. Thus, the proapoptotic role of Bid may involve mitochondrial membrane destabilization, as well as multiple protein-protein interactions.

Specifications

Specifications

Bid
40
Unconjugated
Affinity Purified
Mouse Bid aa. 46-168
50μg
Cell Biology
Primary
Murine
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit