Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biliverdin Reductase Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA1019
Description
PA1-019 detects biliverdin-IX alpha reductase (BLVRA) from human cells. PA1-019 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽40 kDa protein representing BLVRA from HeLa cell extract. The PA1-019 immunogen is purified human biliverdin reductase expressed in E. coli.
Biliverdin reductase (BLVR) is a ubiquitously expressed, small cytoplasmic oxidoreductase that catalyzes the reduction of biliverdin to bilirubin. The first step of this process is catalyzed by heme oxygenase, an enzyme that converts heme to iron, carbon monoxide, and biliverdin. BLVR is unique among all enzymes to date in being dual pH/dual cofactor-dependent, as it requires NADH (pH 6.7) or NADPH (pH 8.7) as an electron donor. There are two isoforms of BLVR, biliverdin-IX alpha-reductase (BLVRA), the major component of human adult liver, and biliverdin–IX beta-reductase (BLVRB), found predomitly in fetal liver. The product of BLVR, bilirubin, is an important biological antioxidant, however it is also a neurotoxin and the cause of kernicterus. In addition to being an antioxidant, it inhibits kinase activity and the activity of enzymes such as protein kinase C and NADPH oxidase.Specifications
Biliverdin Reductase | |
Polyclonal | |
Unconjugated | |
BLVRA | |
BLVRA, Biliverdin reductase A, EC 1.3.1.24, BVR A | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
644 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P53004 | |
BLVRA | |
Purified human biliverdin-IX alpha-reductase expressed in E. coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction