Learn More
anti-Biliverdin Reductase B/BLVRB, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131143
Catalog No. NBP131143
Add to cart
Description
Description
Biliverdin Reductase B/BLVRB Polyclonal antibody specifically detects Biliverdin Reductase B/BLVRB in Human samples. It is validated for Western Blot.
Specifications
Specifications
Biliverdin Reductase B/BLVRB
0.4mg/mL
Western Blot 1:500-1:3000
Affinity Purified
BLVRB
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Flavin reductase. The exact sequence is proprietary.
22 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Rat (85%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Biliverdin reductase B, biliverdin reductase B (flavin reductase (NADPH)), Biliverdin-IX beta-reductase, BVR-B, EC 1.3.1.24, EC 1.5.1.30, flavin reductase, flavin reductase (NADPH), FLRBVRB, FR, GHBP, Green heme-binding protein, MGC117413, NADPH-dependent diaphorase, NADPH-flavin reductase, SDR43U1, short chain dehydrogenase/reductase family 43U, member 1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
645
Human