Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XIAP Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527203

Catalog No. PIPA527203


Add to cart

Description

Description

PA5-27203 targets XIAP in IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-27203 immunogen is recombit fragment corresponding to a region within amino acids 282 and 470 of Human XIAP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7.
Specifications

Specifications

XIAP
Polyclonal
Unconjugated
XIAP
Liquid
XIAP
Recombit fragment corresponding to a region within amino acids 282 and 470 of Human XIAP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.78 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P98170, Q60989
API3, BIRC4, IAP-3, ILP1, MIHA, XLP2, hIAP-3, hIAP3, E3 ubiquitin-protein ligase XIAP, IAP-like protein, X-linked IAP, X-linked inhibitor of apoptosis protein, baculoviral IAP repeat-containing 4, baculoviral IAP repeat-containing protein 4, inhibitor of apoptosis protein 3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11798, 331
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit