Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Livin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14207

Catalog No. PA14207


Add to cart

Description

Description

PA1-4207 detects the Livin protein in human samples. PA1-4207 has been successfully used in Western blot procedures. The PA1-4207 immunogen is a synthetic peptide corresponding to residues G(13) P Q P S H W A A G D G P T Q E R C(30) of human Livin.

Livin is a novel inhibitor of apoptosis protein family members. Disruption of Livin function induces apoptosis in certain cancer cells.
Specifications

Specifications

Livin
Polyclonal
Unconjugated
BIRC7
Liquid
BIRC7
Synthetic peptide corresponding to residues G(13) P Q P S H W A A G D G P T Q E R C(30) of human Livin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96CA5
Baculoviral IAP repeat-containing protein 7, Kidney inhibitor of apoptosis protein, KIAP, Melanoma inhibitor of apoptosis protein, ML-IAP
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79444
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit