Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blimp-1 Rat anti-Mouse, PE-CF594, Clone: 5E7, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564269

Catalog No. BDB564269


Add to cart

Description

Description

The 5E7 monoclonal antibody specifically binds to mouse B lymphocyte-induced maturation protein 1 (Blimp-1). Blimp-1 is a 98 kDa zinc finger-containing protein that is encoded by the Prdm1 gene and functions as a transcriptional repressor. Blimp-1 is essential for regulating the terminal differentiation of activated B cells into plasma cells. In this process, Blimp-1 suppresses genes involved in B cell proliferation, such as Myc and Bcl6 . It controls other genes, such as

This antibody is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Specifications

Specifications

Blimp-1
5.00E+07
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry, Immunocytochemistry
0.2mg/mL
Prdm1; PR domain zinc finger protein 1; Blimp1; PRDI-BF1; ZNFPR1A1
Affinity Purified
Mouse Blimp-1 Recombinant Protein
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit